4-Methyl-2-nitrophenol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Nitro-p-kresol, 1-Hydroxy-4-methyl-2-nitrobenzol, 4-Hydroxy-3-nitrotoluol
A12009.14
A12009.22
A12009.36
A12009.14EA
42.3
EUR
A12009.14
A12009.22
A12009.36
4-Methyl-2-nitrophenol ≥97%
4-Methyl-2-nitrophenol
Purity: 97%
CAS No.: 119-33-5
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Flash Point: 108°(226°F)
Odour: Phenol-like
Boiling Point: 125°/22mm
Beilstein Reference: 1868022
MDL No.: MFCD00007120
Formel:
C₇H₇NO₃ Molecular Weight: 153.14 g/mol Siedepunkt: 125 °C (22 mmHg) Schmelzpunkt: 32…35 °C Dichte: 1.240 Flammpunkt: 108 °C (226 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007120 CAS-Nummer: 119-33-5 EINECS: 204-315-6 UN: 2446 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...