1-Octadecanol ≥97%

Lieferant: Thermo Fisher Scientific

A12020.0E A12020.30 A12020.36
A12020.0EEA 95 EUR
A12020.0E A12020.30 A12020.36
1-Octadecanol ≥97%
1-Octadecanol

Purity: 97% CAS No.: 112-92-5 Molecular Formula: CH3(CH2)17OH Formula Weight: 270.50 Flash Point: 174°(345°F) Physical Form: Flakes or pellets Melting Point: 56-60° Boiling Point: 210°/15mm Beilstein Reference: 1362907 Merck Reference: 14,8805 MDL No.: MFCD00002823

Formel: H₃C(CH₂)₁₆CH₂OH
Siedepunkt: 210 °C (15 mmHg)
Schmelzpunkt: 56…60 °C
Dichte: 0,805…0.815 g/cm³ (60 °C)
Flammpunkt: 174 °C (345 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002823
CAS-Nummer: 112-92-5
EINECS: 204-017-6
Merck Index: 14,8805

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR