1-Octadecanol ≥97%
Lieferant: Thermo Fisher Scientific
Purity: 97%
CAS No.: 112-92-5
Molecular Formula: CH3(CH2)17OH
Formula Weight: 270.50
Flash Point: 174°(345°F)
Physical Form: Flakes or pellets
Melting Point: 56-60°
Boiling Point: 210°/15mm
Beilstein Reference: 1362907
Merck Reference: 14,8805
MDL No.: MFCD00002823
Formel:
H₃C(CH₂)₁₆CH₂OH Siedepunkt: 210 °C (15 mmHg) Schmelzpunkt: 56…60 °C Dichte: 0,805…0.815 g/cm³ (60 °C) Flammpunkt: 174 °C (345 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002823 CAS-Nummer: 112-92-5 EINECS: 204-017-6 Merck Index: 14,8805 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...