Ethyltiglat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12029.14 A12029.22
A12029.14EA 40.8 EUR
A12029.14 A12029.22
Ethyltiglat ≥98%
Ethyltiglat

Purity: 98% CAS No.: 5837-78-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 46°(114°F) Refractive Index: 1.4340 Boiling Point: 156° Beilstein Reference: 1720895 Merck Reference: 14,9433 UN No.: 3272 MDL No.: MFCD00015183

Formel: C₇H₁₂O₂
Siedepunkt: 156 °C
Dichte: 0,930
Flammpunkt: 46 °C (114 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015183
CAS-Nummer: 5837-78-5
EINECS: 227-425-6
UN: 3272
ADR: 3,III
Merck Index: 14,9433

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR