Chloramin T (Natriumsalz) Trihydrat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chloramin T Natriumsalz Trihydrat, Tosylchloramidnatrium Trihydrat, N-Chlor-p-toluolsulfonamid Natriumsalz Trihydrat

A12044.0B A12044.0I A12044.30
A12044.0BEA 91.1 EUR
A12044.0B A12044.0I A12044.30
Chloramin T (Natriumsalz) Trihydrat 98%
Chloramin T (Natriumsalz) Trihydrat

Purity: 98% CAS No.: 7080-50-4 Molecular Formula: C7H7ClNNaO2S.3H2O Formula Weight: 281.69 (227.67anhy) Flash Point: 192°(377°F) Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White to pale yellow Odour: Chlorine-like pH: 8-10 Melting Point: 167-170° Beilstein Reference: 3924168 Merck Reference: 14,2075 UN No.: 3263 MDL No.: MFCD00149066

Formel: CH₃C₆H₄SO₂NClNa·3H₂O
Molecular Weight: 281.69 g/mol
Schmelzpunkt: 167…170 °C
Dichte: 1.43 g/cm³ (20 °C)
Flammpunkt: 192 °C (377 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149066
CAS-Nummer: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8 III
Merck Index: 13,02085

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR