4-Ethylbrenzcatechin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Ethylbenzol-1,2-diol

A12048.03 A12048.06 A12048.MD
A12048.03EA 83.1 EUR
A12048.03 A12048.06 A12048.MD
4-Ethylbrenzcatechin ≥98%
4-Ethylbrenzcatechin

Purity: 98% CAS No.: 1124-39-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 37-40° Boiling Point: 105-107°/0.5mm MDL No.: MFCD00015847

Formel: C₈H₁₀O₂
Molecular Weight: 138.17 g/mol
Siedepunkt: 105…107 °C (0.5 mmHg)
Schmelzpunkt: 37…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015847
CAS-Nummer: 1124-39-6
EINECS: 214-397-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR