(±)-1-Octen-3-ol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12079.09
A12079.18
A12079.30
A12079.09EA
29.2
EUR
A12079.09
A12079.18
A12079.30
(±)-1-Octen-3-ol ≥98%
(±)-1-Octen-3-ol
Purity: 98%
CAS No.: 3391-86-4
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 61°(141°F)
Refractive Index: 1.4370
Odour: Characteristic
Melting Point: -49°
Boiling Point: 173-175°
Beilstein Reference: 1744110
MDL No.: MFCD00004589
Formel:
C₈H₁₆O Siedepunkt: 173…175 °C Schmelzpunkt: –49 °C Dichte: 0,833 Flammpunkt: 61 °C (141 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004589 CAS-Nummer: 3391-86-4 EINECS: 222-226-0 UN: 2810 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...