trans,trans-Dibenzylidenaceton ≥98%

Lieferant: Thermo Fisher Scientific

A12095.06 A12095.14 A12095.22
A12095.06EA 38.9 EUR
A12095.06 A12095.14 A12095.22
trans,trans-Dibenzylidenaceton ≥98%
trans,trans-Dibenzylidenaceton

Purity: 98+% CAS No.: 35225-79-7 Molecular Formula: C17H14O Formula Weight: 234.30 Melting Point: 112-114° Beilstein Reference: 608434 Merck Reference: 14,3006 MDL No.: MFCD00004790

Formel: C₁₇H₁₄O
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004790
CAS-Nummer: 35225-79-7
Merck Index: 14,3006

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR