L(+)-Ornithin Hydrochlorid ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

A12111.14 A12111.22 A12111.36
A12111.14EA 28.3 EUR
A12111.14 A12111.22 A12111.36
L(+)-Ornithin Hydrochlorid ≥99%
L(+)-Ornithin Hydrochlorid

Purity: 99% CAS No.: 3184-13-2 Molecular Formula: C5H12N2O2.HCl Formula Weight: 168.62 Melting Point: ca 238° dec. Beilstein Reference: 3625847 Merck Reference: 14,6874 Optical Rotation: +23° (c=5 in 5N HCl) MDL No.: MFCD00064562

Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168.62 g/mol
Schmelzpunkt: ∼ 238 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR