Kaliumdihydrogenphosphat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12142.0E A12142.36
A12142.0EEA 139 EUR
A12142.0E A12142.36
Kaliumdihydrogenphosphat ≥98%
Kaliumdihydrogenphosphat

Purity: 98+% CAS No.: 7778-77-0 Molecular Formula: H2KO4P Formula Weight: 136.09 Physical Form: Cryst solid Density: 2.338 Appearance: White Melting Point: 253° Merck Reference: 14,7659 MDL No.: MFCD00011401

Formel: KH₂PO₄
Siedepunkt: ∼100 °C (1013 hPa)
Schmelzpunkt: 253 °C
Dichte: 2,338
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 14,7659

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR