trans-Indole-3-acrylic acid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12143.03
A12143.06
A12143.03EA
48.2
EUR
A12143.03
A12143.06
trans-Indole-3-acrylic acid ≥98%
trans-Indole-3-acrylic acid
Purity: 98+%
CAS No.: 29953-71-7
Molecular Formula: C11H9NO2
Formula Weight: 187.20
Storage Temperature: 2°C to 8°C
Melting Point: ca 185°dec.
Beilstein Reference: 6317
MDL No.: MFCD00005633
Formel:
C₁₁H₉NO₂ |
MDL:
MFCD00005633 CAS-Nummer: 29953-71-7 EINECS: 214-872-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...