Methylviolett 2B

Lieferant: Thermo Fisher Scientific
Achtung

A12159.18 A12159.30
A12159.18EA 30.9 EUR
A12159.18 A12159.30
Methylviolett 2B
Methylviolett 2B

CAS No.: 8004-87-3 Molecular Formula: C24H27N3.HCl Formula Weight: 393.96 Odour: Odourless MDL No.: MFCD00012568

Formel: C₂₄H₂₈ClN₃
Schmelzpunkt: 137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012568
CAS-Nummer: 8004-87-3
UN: 3077
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR