N-(2-Bromethyl)phthalimid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(2-Bromethyl)phthalsäureimid, N-(2-Bromethyl)-1,2-benzoldicarbonsäureimid

A12164.14 A12164.22 A12164.36
A12164.14EA 33 EUR
A12164.14 A12164.22 A12164.36
N-(2-Bromethyl)phthalimid ≥98%
N-(2-Bromethyl)phthalimid

Purity: 98+% CAS No.: 574-98-1 Molecular Formula: C10H8BrNO2 Formula Weight: 254.09 Melting Point: 80-84° Beilstein Reference: 148736

Formel: C₁₀H₈BrNO₂
Siedepunkt: 318 °C (1013 hPa)
Schmelzpunkt: 80…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005902
CAS-Nummer: 574-98-1
EINECS: 209-379-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR