Sudan IV

Lieferant: Thermo Fisher Scientific
Achtung

A12181.22 A12181.14
A12181.22EA 64.2 EUR
A12181.22 A12181.14
Sudan IV
Sudan IV

CAS No.: 85-83-6 Molecular Formula: C24H20N4O Formula Weight: 380.45 Melting Point: ca 185° dec. Merck Reference: 14,8393 MDL No.: MFCD00003893

Formel: C₂₄H₂₀N₄O
Schmelzpunkt: ∼ 185 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003893
CAS-Nummer: 85-83-6
EINECS: 201-635-8
Merck Index: 14,8393

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR