2-Chlorbenzimidazol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chlor-1H-benzimidazol
A12193.06
A12193.14
A12193.22
A12193.06EA
67.3
EUR
A12193.06
A12193.14
A12193.22
2-Chlorbenzimidazol ≥97%
2-Chlorobenzimidazole
Purity: 97%
CAS No.: 4857-06-1
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: ca 180-185°
Beilstein Reference: 116526
Formel:
C₇H₅ClN₂ Molecular Weight: 152.58 g/mol Schmelzpunkt: ∼ 180…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051944 CAS-Nummer: 4857-06-1 EINECS: 225-453-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...