2-Chlorbenzimidazol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chlor-1H-benzimidazol

A12193.06 A12193.14 A12193.22
A12193.06EA 67.3 EUR
A12193.06 A12193.14 A12193.22
2-Chlorbenzimidazol ≥97%
2-Chlorobenzimidazole

Purity: 97% CAS No.: 4857-06-1 Molecular Formula: C7H5ClN2 Formula Weight: 152.58 Melting Point: ca 180-185° Beilstein Reference: 116526

Formel: C₇H₅ClN₂
Molecular Weight: 152.58 g/mol
Schmelzpunkt: ∼ 180…185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051944
CAS-Nummer: 4857-06-1
EINECS: 225-453-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR