2,6-Dimethoxybenzaldehyd ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12209.03
A12209.06
A12209.14
A12209.03EA
50.5
EUR
A12209.03
A12209.06
A12209.14
2,6-Dimethoxybenzaldehyd ≥99%
2,6-Dimethoxybenzaldehyde
Purity: 99%
CAS No.: 3392-97-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 96-98°
Boiling Point: 285°
MDL No.: MFCD00010862
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 285 °C Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010862 CAS-Nummer: 3392-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...