2-Chlor-4-(trifluormethyl)anilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12242.06
A12242.14
A12242.22
A12242.06EA
50.1
EUR
A12242.06
A12242.14
A12242.22
2-Chlor-4-(trifluormethyl)anilin ≥98%
2-Chlor-4-(trifluormethyl)anilin
Purity: 98%
CAS No.: 39885-50-2
Molecular Formula: C7H5ClF3N
Formula Weight: 195.57
Flash Point: 103°(217°F)
Refractive Index: 1.5015
Boiling Point: 214-218°
Beilstein Reference: 4179703
UN No.: 2810
MDL No.: MFCD00042563
Formel:
C₇H₅ClF₃N Siedepunkt: 214…218 °C Dichte: 1,434 Flammpunkt: 103 °C (217 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042563 CAS-Nummer: 39885-50-2 EINECS: 254-674-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...