3,4-Dichlorphenol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12256.14
A12256.22
A12256.14EA
78
EUR
A12256.14
A12256.22
3,4-Dichlorphenol ≥99%
3,4-Dichlorphenol
Purity: 99%
CAS No.: 95-77-2
Molecular Formula: C6H4Cl2O
Formula Weight: 163.00
Odour: Phenol-like
Melting Point: 64-68°
Boiling Point: 252-255°
Beilstein Reference: 1907693
UN No.: 2020
MDL No.: MFCD00002258
Formel:
C₆H₄Cl₂O Siedepunkt: 252…255 °C Schmelzpunkt: 64…68 °C Dichte: 1,6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002258 CAS-Nummer: 95-77-2 EINECS: 202-450-5 UN: 2020 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...