Dichloromethylenedimethyliminium chloride 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12300.14
A12300.22
A12300.14EA
87.5
EUR
A12300.14
A12300.22
Dichloromethylenedimethyliminium chloride 95%
Dichloromethylenedimethyliminium chloride
Purity: 95%
CAS No.: 33842-02-3
Molecular Formula: C3H6Cl3N
Formula Weight: 162.45
Storage Temperature: 2°C to 8°C
Melting Point: ca 185° dec.
Beilstein Reference: 4157987
UN No.: UN3261
MDL No.: MFCD00011870
Formel:
C₃H₆Cl₃N |
MDL:
MFCD00011870 CAS-Nummer: 33842-02-3 EINECS: 251-695-4 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...