Ethyl 3-Methylpyrazole-5-carboxylate 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Ethyl 3-methyl-1H-pyrazole-5-carboxylate

A12304.06 A12304.14
A12304.06EA 132 EUR
A12304.06 A12304.14
Ethyl 3-Methylpyrazole-5-carboxylate 97%
Ethyl 3-Methylpyrazole-5-carboxylate

Purity: 97% CAS No.: 4027-57-0 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Melting Point: 80-83° Beilstein Reference: 4192 MDL No.: MFCD00052514

Formel: C₇H₁₀N₂O₂
Schmelzpunkt: 80…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052514
CAS-Nummer: 4027-57-0
EINECS: 223-702-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR