Natriumchlorid 99+%, kristallines Pulver PDV

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Saline, NaCl, Sea salt, Halite

A12313.0B A12313.0G A12313.0I
A12313.0BEA 24 EUR
A12313.0B A12313.0G A12313.0I
Natriumchlorid 99+%, kristallines Pulver PDV
Natriumchlorid

Purity: 99+% CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Refractive Index: 1.5442 Odour: Odourless pH: 6-9 Melting Point: 801° Boiling Point: 1413° Merck Reference: 14,8599 MDL No.: MFCD00003477

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1413 °C
Schmelzpunkt: 801
Dichte: 2.165
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,8599

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR