Benzo[b]thiophen ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12328.06 A12328.14 A12328.22
A12328.06EA 36.2 EUR
A12328.06 A12328.14 A12328.22
Benzo[b]thiophen ≥98%
Benzo[b]thiophen

Purity: 98+% CAS No.: 95-15-8 Molecular Formula: C8H6S Formula Weight: 134.20 Flash Point: 91°(195°F) Odour: Unpleasant Boiling Point: 221-222° Beilstein Reference: 80580 Merck Reference: 14,9303 UN No.: 3077 MDL No.: MFCD00005864

Formel: C₈H₆S
Siedepunkt: 221…222 °C
Schmelzpunkt: 28…32 °C
Dichte: 1,149
Flammpunkt: 91 °C (195 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005864
CAS-Nummer: 95-15-8
EINECS: 202-395-7
ADR: 9,III
Merck Index: 14,9303

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR