2,2'-Bithiophen ≥98%

Lieferant: Thermo Fisher Scientific

A12335.03 A12335.06 A12335.14
A12335.03EA 36 EUR
A12335.03 A12335.06 A12335.14
2,2'-Bithiophen ≥98%
2,2'-Bithiophen

Purity: 98% CAS No.: 492-97-7 Molecular Formula: C8H6S2 Formula Weight: 166.27 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 260° Beilstein Reference: 3039 MDL No.: MFCD00005414

Formel: C₈H₆S₂
Siedepunkt: 260 °C
Schmelzpunkt: 30…33 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005414
CAS-Nummer: 492-97-7
EINECS: 207-767-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR