Calciumdibenzoat Hydrat, (max. 8% H₂O) 98%

Lieferant: Thermo Fisher Scientific

A12358.36 A12358.22
A12358.36EA 149 EUR
A12358.36 A12358.22
Calciumdibenzoat Hydrat, (max. 8% H₂O) 98%
Calciumdibenzoat Hydrat

Calcium benzoate hydrate is used in used in the food industry as a preservative.


Purity: 98% CAS No.: 2090-05-3 Molecular Formula: (C6H5CO2)2Ca.xH2O Formula Weight: 282.32(anhy) MDL No.: MFCD00167164

Formel: C₁₄H₁₀CaO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00167164
CAS-Nummer: 2090-05-3
EINECS: 218-235-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR