Diphenylcarbonat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12362.0B
A12362.0I
A12362.30
A12362.0BEA
65.6
EUR
A12362.0B
A12362.0I
A12362.30
Diphenylcarbonat ≥99%
Diphenylcarbonat
Purity: 99%
CAS No.: 102-09-0
Molecular Formula: C13H10O3
Formula Weight: 214.22
Flash Point: 168°(334°F)
Odour: Mild
Boiling Point: 301-302°
Beilstein Reference: 1074863
Merck Reference: 14,3602
MDL No.: MFCD00003037
Formel:
C₁₃H₁₀O₃ Siedepunkt: 301…302 °C Schmelzpunkt: 76…81 °C Dichte: 1,300 Flammpunkt: 168 °C (334 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003037 CAS-Nummer: 102-09-0 EINECS: 203-005-8 Merck Index: 14,3602 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...