Trimethyl-4-phosphonocrotonate Gemisch aus EZ-Isomeren ≥90%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12399.09
A12399.18
A12399.09EA
85.5
EUR
A12399.09
A12399.18
Trimethyl-4-phosphonocrotonate Gemisch aus EZ-Isomeren ≥90%
Trimethyl-4-phosphonocrotonate
Purity: 90+%
CAS No.: 86120-40-3
Molecular Formula: C7H13O5P
Formula Weight: 208.15
Boiling Point: 123-124°/1mm
Beilstein Reference: 4987133
MDL No.: MFCD00051564
Formel:
C₇H₁₃O₅P Siedepunkt: 123…124 °C (1 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051564 CAS-Nummer: 86120-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...