Taurin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A12403.0E A12403.22 A12403.36
A12403.0EEA 363 EUR
A12403.0E A12403.22 A12403.36
Taurin ≥99%
Taurin

Purity: 99% CAS No.: 107-35-7 Molecular Formula: H2NCH2CH2SO3H Formula Weight: 125.15 Physical Form: Crystals or cryst powder Appearance: White Melting Point: 300° Beilstein Reference: 1751215 Merck Reference: 14,9074 MDL No.: MFCD00008197

Formel: C₂H₇NO₃S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008197
CAS-Nummer: 107-35-7
EINECS: 203-483-8
Merck Index: 14,9074

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR