2-Brombenzylamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2-Bromophenyl)methanamine

A12412.14 A12412.06
A12412.14EA 242 EUR
A12412.14 A12412.06
2-Brombenzylamin 97%
2-Brombenzylamin

Purity: 98% CAS No.: 3959-05-5 Molecular Formula: C7H8BrN Formula Weight: 186.06 Odour: Amine-like Boiling Point: 118-119°/9mm UN No.: 2735 MDL No.: MFCD00025572

Formel: C₇H₈BrN
Molecular Weight: 186.05 g/mol
Siedepunkt: 118…119 °C (9 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00025572
CAS-Nummer: 3959-05-5
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR