Piperidin ≥99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 110-89-4
Molecular Formula: C5H11N
Formula Weight: 85.15
Flash Point: 3°(37°F)
Physical Form: Liquid
Density: 0.862
Refractive Index: 1.4520
Sensitivity: Air Sensitive
Appearance: Colourless
Melting Point: -11°
Boiling Point: 105-106°
Beilstein Reference: 102438
Merck Reference: 14,7468
UN No.: 2401
MDL No.: MFCD00005979
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...