1,8-Diazabicyclo[5.4.0]-7-undecen ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 6674-22-2
Molecular Formula: C9H16N2
Formula Weight: 152.24
Flash Point: 116°(240°F)
Refractive Index: 1.5220
Odour: Amine-like
pH: 12.8
Melting Point: -70°
Boiling Point: 261°
Beilstein Reference: 508906
UN No.: 3267
MDL No.: MFCD00006930
Formel:
C₉H₁₆N₂ Siedepunkt: 261 °C Schmelzpunkt: –70 °C Dichte: 1,020 Flammpunkt: 116 °C (240 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006930 CAS-Nummer: 6674-22-2 EINECS: 229-713-7 UN: 3267 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...