(2,4-Dichlorphenoxy)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A12467.0B A12467.18 A12467.30
A12467.0BEA 311 EUR
A12467.0B A12467.18 A12467.30
(2,4-Dichlorphenoxy)essigsäure ≥98%
(2,4-Dichlorphenoxy)essigsäure

Purity: 98% CAS No.: 94-75-7 Molecular Formula: C8H6Cl2O3 Formula Weight: 221.04 Odour: Odourless Melting Point: 136-140° Boiling Point: 160°/0.4mm Beilstein Reference: 1214242 Merck Reference: 14,2796 MDL No.: MFCD00004300

Formel: (C₆H₃Cl₂)OCH₂COOH
Siedepunkt: 160 °C (0,4 mmHg)
Schmelzpunkt: 136…140 °C
Dichte: 1,42
Lagertemperatur: Raumtemperatur
MDL: MFCD00004300
CAS-Nummer: 94-75-7
EINECS: 202-361-1
Merck Index: 14,2796

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR