Ammoniumeisen(II)sulfat Hexahydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ferroammoniumsulfat Hexahydrat, Eisen(II)ammoniumsulfat Hexahydrat, Mohrsches Salz
A12473.36
A12473.36EA
54.8
EUR
A12473.36
Ammoniumeisen(II)sulfat Hexahydrat ≥99%
Ammoniumeisen(II)sulfat Hexahydrat
Purity: 99%
CAS No.: 7783-85-9
Molecular Formula: H8FeN2O8S2.6H2O
Formula Weight: 392.13 (284.04anhy)
Odour: Odourless
Melting Point: ca 100° dec.
MDL No.: MFCD00150530
Formel:
(NH₄)₂Fe(SO₄)₂·6H₂O Molecular Weight: 392.14 g/mol Schmelzpunkt: ∼ 100 °C (Zers.) Dichte: 1.864 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150530 CAS-Nummer: 7783-85-9 EINECS: 233-382-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...