5-(Hydroxymethyl)-2-furaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12475.03
A12475.06
A12475.14
A12475.03EA
62.2
EUR
A12475.03
A12475.06
A12475.14
5-(Hydroxymethyl)-2-furaldehyd ≥97%
5-(Hydroxymethyl)-2-furaldehyd
Purity: 97%
CAS No.: 67-47-0
Molecular Formula: C6H6O3
Formula Weight: 126.11
Storage Temperature: 2°C to 8°C
Flash Point: 79°(174°F)
Refractive Index: 1.5620
Melting Point: 32-35°
Boiling Point: 114-116°/1mm
Beilstein Reference: 110889
Merck Reference: 14,4832
MDL No.: MFCD00003234
Formel:
C₆H₆O₃ Siedepunkt: 114…116 °C (1 mmHg) Schmelzpunkt: 32…35 °C Dichte: 1,206 Flammpunkt: 79 °C (174 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00003234 CAS-Nummer: 67-47-0 EINECS: 200-654-9 Merck Index: 14,4832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...