Quecksilber(II)acetat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A12478.14 A12478.22 A12478.36
A12478.14EA 32 EUR
A12478.14 A12478.22 A12478.36
Quecksilber(II)acetat ≥98%

Purity: 98+% CAS No.: 1600-27-7 Molecular Formula: C4H6HgO4 Formula Weight: 318.68 Melting Point: 179-182° Beilstein Reference: 3563831 Merck Reference: 14,5873 UN No.: 1629 MDL No.: MFCD00012165

Formel: (H₃CCOO)₂Hg
Schmelzpunkt: 179…182 °C
Dichte: 3,28
Lagertemperatur: Raumtemperatur
MDL: MFCD00012165
CAS-Nummer: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 14,5873

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR