Tris(diethylamino)phosphin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Hexaethylphosphortriamid

A12490.06 A12490.14 A12490.22
A12490.06EA 50.2 EUR
A12490.06 A12490.14 A12490.22
Tris(diethylamino)phosphin ≥97%
Tris(diethylamino)phosphin

Purity: 97% CAS No.: 2283-11-6 Molecular Formula: [(CH3CH2)2N]3P Formula Weight: 247.37 Flash Point: 58°(136°F) Refractive Index: 1.4737 Boiling Point: 245-246° Beilstein Reference: 636187 UN No.: 1993 MDL No.: MFCD00009041

Formel: C₁₂H₃₀N₃P
Molecular Weight: 247.36 g/mol
Siedepunkt: 245…246 °C
Dichte: 0.903
Flammpunkt: 58 °C (136 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009041
CAS-Nummer: 2283-11-6
EINECS: 218-920-8
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR