N,N,N',N'-Tetramethylethylendiamin (TEMED) 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12536.0F
A12536.AE
A12536.AP
A12536.0FEA
282
EUR
A12536.0F
A12536.AE
A12536.AP
N,N,N',N'-Tetramethylethylendiamin (TEMED) 99%
N,N,N',N'-Tetramethylethylendiamin (TEMED)
Purity: 99%
CAS No.: 110-18-9
Molecular Formula: C6H16N2
Formula Weight: 116.21
Flash Point: 18°(62°F)
Physical Form: Liquid
Density: 0.775
Refractive Index: 1.4180
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless to pale yellow
Odour: Unpleasant
Melting Point: -55°
Boiling Point: 120-122°
Beilstein Reference: 1732991
UN No.: 2372
MDL No.: MFCD00008335
Formel:
C₆H₁₆N₂ Siedepunkt: 120…122 °C Schmelzpunkt: –55 °C Dichte: 0,775 Flammpunkt: 18 °C (62 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008335 CAS-Nummer: 110-18-9 EINECS: 203-744-6 UN: 2372 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...