ω-Brom-4-chloracetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A12607.14 A12607.22 A12607.36
A12607.14EA 47.1 EUR
A12607.14 A12607.22 A12607.36
ω-Brom-4-chloracetophenon ≥98%
ω-Brom-4-chloracetophenon

Purity: 98% CAS No.: 536-38-9 Molecular Formula: C8H6BrClO Formula Weight: 233.50 Melting Point: 94-98° Beilstein Reference: 607603 Merck Reference: 14,2153 UN No.: 3261 MDL No.: MFCD00000625

Formel: C₈H₆BrClO
Siedepunkt: 278 °C (1013 hPa)
Schmelzpunkt: 94…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000625
CAS-Nummer: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: 8,III
Merck Index: 14,2153

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR