Pyrrol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1H-Pyrrol
A12616.30
A12616.18
A12616.0B
A12616.30EA
137
EUR
A12616.30
A12616.18
A12616.0B
Pyrrol ≥98%
Pyrrol
Purity: 98+%
CAS No.: 109-97-7
Molecular Formula: C4H5N
Formula Weight: 67.09
Storage Temperature: 2°C to 8°C
Flash Point: 38°(100°F)
Refractive Index: 1.5090
Odour: Mild
Boiling Point: 131°
Beilstein Reference: 1159
Merck Reference: 14,8014
UN No.: 1992
MDL No.: MFCD00005216
Formel:
C₄H₅N Siedepunkt: 131 °C Schmelzpunkt: –24 °C Dichte: 0,967 Flammpunkt: 38 °C (100 °F) |
MDL:
MFCD00005216 CAS-Nummer: 109-97-7 EINECS: 203-724-7 UN: 1992 ADR: 3,III Merck Index: 14,8014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...