Butyrylthiocholiniodid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Butyrylthiocholiniumiodid, (Propylcarbonylthioethyl)trimethylammoniumiodid, (2-(Butyrylthio)ethyl)trimethylammoniumiodid, (2-Butyrylthioethyl)trimethylammoniumiodid

A12622.06 A12622.14
A12622.06EA 61.5 EUR
A12622.06 A12622.14
Butyrylthiocholiniodid ≥98%
Butyrylthiocholiniodid

Purity: 98% CAS No.: 1866-16-6 Molecular Formula: C9H20INOS Formula Weight: 317.23 Storage Temperature: 2°C to 8°C Melting Point: 172-176° Beilstein Reference: 3729509 MDL No.: MFCD00011845

Formel: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
Molecular Weight: 317,23 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011845
CAS-Nummer: 1866-16-6
EINECS: 217-475-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR