3-Methoxybenzylchlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 824-98-6
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Storage Temperature: 2°C to 8°C
Flash Point: 101°(213°F)
Refractive Index: 1.5440
Boiling Point: 122-123°/12mm
Beilstein Reference: 636684
UN No.: 1760
MDL No.: MFCD00000907
Formel:
C₈H₉ClO Siedepunkt: 122…123 °C (12 mmHg) Dichte: 1,078 Flammpunkt: 101 °C (213 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000907 CAS-Nummer: 824-98-6 EINECS: 212-541-1 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...