(S)-(-)-α-Methylbenzylamin 98%
Lieferant: Thermo Fisher Scientific
Purity: 98%
CAS No.: 2627-86-3
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 71°(159°F)
Refractive Index: 1.5260
Odour: Amine-like
Melting Point: -10°
Boiling Point: 187°
Beilstein Reference: 2204907
Merck Reference: 14,6026
Optical Rotation: -30° (c=10 in ethanol)
UN No.: 2735
MDL No.: MFCD00064406
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...