Trimethylsilylisocyanat ≥94%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 94%
CAS No.: 1118-02-1
Molecular Formula: C4H9NOSi
Formula Weight: 115.21
Storage Temperature: -30°C to -10°C
Flash Point: -2°(28°F)
Refractive Index: 1.3960
Odour: Acrid
Melting Point: -49°
Boiling Point: 90-92°
Beilstein Reference: 1744962
UN No.: 2478
MDL No.: MFCD00001993
Formel:
C₄H₉NOSi Siedepunkt: 90…92 °C Schmelzpunkt: –49 °C Dichte: 0,859 Flammpunkt: –2 °C (28 °F) |
MDL:
MFCD00001993 CAS-Nummer: 1118-02-1 EINECS: 214-256-8 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...