3-Indolcarbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1H-Indole-3-carboxylic acid

A12634.03 A12634.06 A12634.14 AB112970.0025
A12634.03EA 36.1 EUR
A12634.03 A12634.06 A12634.14 ABCRAB112970.0025
3-Indolcarbonsäure ≥98%
3-Indolcarbonsäure

Purity: 98% CAS No.: 771-50-6 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: ca 239° dec. Beilstein Reference: 129435

Formel: C₉H₇NO₂
Schmelzpunkt: ∼ 239 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005624
CAS-Nummer: 771-50-6
EINECS: 212-231-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR