2-Hexylthiophen ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12652.09 A12652.18
A12652.09EA 74.3 EUR
A12652.09 A12652.18
2-Hexylthiophen ≥98%
2-Hexylthiophen

Purity: 98% CAS No.: 18794-77-9 Molecular Formula: C10H16S Formula Weight: 168.30 Flash Point: 100°(212°F) Refractive Index: 1.4960 Boiling Point: 228-230° Beilstein Reference: 110220 MDL No.: MFCD00022535

Formel: C₁₀H₁₆S
Siedepunkt: 228…230 °C
Dichte: 0,935
Flammpunkt: 100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00022535
CAS-Nummer: 18794-77-9
EINECS: 242-579-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR