α-Chlor-3-fluortoluol 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: α-Chlor-m-fluortoluol

A12665.14 A12665.06 A12665.22
A12665.14EA 119 EUR
A12665.14 A12665.06 A12665.22
α-Chlor-3-fluortoluol 97%
α-Chlor-3-fluortoluol

Purity: 97% CAS No.: 456-42-8 Molecular Formula: C7H6ClF Formula Weight: 144.58 Flash Point: 58°(136°F) Refractive Index: 1.5120 Odour: Pungent Boiling Point: 176-177° Beilstein Reference: 742265 UN No.: 2920 MDL No.: MFCD00000904

Formel: C₇H₆ClF
Molecular Weight: 144.58 g/mol
Siedepunkt: 176…177 °C
Dichte: 1.213
Flammpunkt: 58 °C (136 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000904
CAS-Nummer: 456-42-8
EINECS: 207-264-8
UN: 2920
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR