4-Aminobenzoesäure 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Carbonsäureanilin, PABA

A12673.0B A12673.18 A12673.30 A12673.PG
A12673.0BEA 144 EUR
A12673.0B A12673.18 A12673.30 A12673.PG
4-Aminobenzoesäure 99%
4-Aminobenzoesäure

Purity: 99% CAS No.: 150-13-0 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: ca 187° dec. Beilstein Reference: 471605 Merck Reference: 14,423 MDL No.: MFCD00007894

Formel: C₇H₇NO₂
Siedepunkt: 340 °C (1013 hPa)
Schmelzpunkt: ∼187 °C (Zers.)
Dichte: 1,374
Lagertemperatur: Raumtemperatur
MDL: MFCD00007894
CAS-Nummer: 150-13-0
EINECS: 205-753-0
Merck Index: 14,423

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR