Methyl-2-thiophencarboxylat 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Methyl thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Methyl Ester
A12677.14
A12677.06
A12677.22
A12677.14EA
183
EUR
A12677.14
A12677.06
A12677.22
Methyl-2-thiophencarboxylat 97%
Methyl-2-thiophencarboxylat
Purity: 97%
CAS No.: 5380-42-7
Molecular Formula: C6H6O2S
Formula Weight: 142.18
Refractive Index: 1.5420
Boiling Point: 95-97°/15mm
Beilstein Reference: 111111
MDL No.: MFCD00016895
Formel:
C₆H₆O₂S Molecular Weight: 142.17 g/mol Siedepunkt: 95…97 °C (15 mmHg) Dichte: 1.230 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016895 CAS-Nummer: 5380-42-7 EINECS: 226-371-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...