Nicotinsäure (Niacin) ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Picolinsäure, Vitamin B3, 3-Pyridincarbonsäure, Pyridin-3-carbonsäure, Niacin

A12683.30 A12683.0B A12683.0I
A12683.30EA 35.1 EUR
A12683.30 A12683.0B A12683.0I
Nicotinsäure (Niacin) ≥99%
Nicotinsäure (Niacin)

Purity: 99% CAS No.: 59-67-6 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 293°(559°F) Physical Form: Powder Density: 1.4 Appearance: White Odour: Odourless pH: 2.7 Melting Point: 236-239° Beilstein Reference: 109591 Merck Reference: 14,6525 MDL No.: MFCD00006391


Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP

Formel: C₆H₅NO₂
Molecular Weight: 123.11 g/mol
Siedepunkt: 258
Schmelzpunkt: 236…239 °C
Dichte: 1.4
Flammpunkt: 293 °C (559 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006391
CAS-Nummer: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR