Ethyl cyano(hydroxyimino)acetate 97%

Lieferant: Thermo Fisher Scientific
Gefahr

A12691.06 A12691.14 A12691.22
A12691.06EA 27.6 EUR
A12691.06 A12691.14 A12691.22
Ethyl cyano(hydroxyimino)acetate 97%
Ethyl cyano(hydroxyimino)acetate

Purity: 97% CAS No.: 3849-21-6 Molecular Formula: C5H6N2O3 Formula Weight: 142.11 Melting Point: 127-132° MDL No.: MFCD00000625

Formel: C₅H₆N₂O₃
Schmelzpunkt: 127…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002112
CAS-Nummer: 3849-21-6
EINECS: 223-351-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR